Assay Details
Gene Symbol: Trap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026508.2 | NP_080784.1 | 4 - 5 | 506 | 70 | |
GenBank mRNA | AK002409.1 | - | 4 - 5 | 492 | 70 | |
AK010341.1 | - | 4 - 5 | 506 | 70 | ||
AK088471.1 | - | 4 - 5 | 495 | 70 | ||
AK143494.1 | - | 4 - 5 | 524 | 70 | ||
AK162067.1 | - | 4 - 5 | 494 | 70 | ||
AK167181.1 | - | 4 - 5 | 495 | 70 | ||
AK188761.1 | - | 1 - 2 | 80 | 70 | ||
BC006685.1 | - | 4 - 5 | 474 | 70 | ||
BC006864.1 | - | 4 - 5 | 474 | 70 | ||
BC022912.1 | - | 4 - 5 | 503 | 4242394 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
