Assay Details
Gene Symbol: Coasy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF521095.1 | - | 3 - 4 | 952 | 125 | |
AK004878.1 | - | 3 - 4 | 1012 | 125 | ||
AK159405.1 | - | 3 - 4 | 1001 | 125 | ||
AK159912.1 | - | 2 - 3 | 1301 | 125 | ||
AK166894.1 | - | 2 - 3 | 1295 | 125 | ||
AK170330.1 | - | 2 - 3 | 1310 | 125 | ||
BC020046.1 | - | 2 - 3 | 422 | 125 | ||
BC064753.1 | - | 2 - 3 | 940 | 125 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC020046.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
