Assay Details
Gene Symbol: Copb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033370.3 | NP_203534.1 | 8 - 9 | 1088 | 98 | |
GenBank mRNA | AF231925.1 | - | 7 - 8 | 1027 | 98 | |
AK011453.1 | - | 8 - 9 | 1112 | 98 | ||
AK135168.1 | - | 8 - 9 | 1120 | 98 | ||
AK149828.1 | - | 8 - 9 | 1106 | 98 | ||
AK152222.1 | - | 8 - 9 | 1115 | 98 | ||
AK161544.1 | - | 8 - 9 | 1115 | 98 | ||
AK172214.1 | - | 8 - 9 | 1090 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
