Assay Details
Gene Symbol: Tnfaip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009396.2 | NP_033422.2 | 9 - 10 | 1698 | 85 | |
GenBank mRNA | AK087680.1 | - | 8 - 9 | 2360 | 85 | |
AK154155.1 | - | 9 - 10 | 1752 | 85 | ||
AK154405.1 | - | 9 - 10 | 1698 | 85 | ||
AK172653.1 | - | 9 - 10 | 1698 | 85 | ||
BC008165.1 | - | 2 - 3 | 196 | 85 | ||
BC138106.1 | - | 8 - 9 | 1607 | 8861125 | 85 | |
BC138107.1 | - | 8 - 9 | 1607 | 8861126 | 85 | |
L24118.1 | - | 8 - 9 | 1642 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
