Assay Details
Gene Symbol: Nbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013752.3 | NP_038780.3 | 9 - 10 | 1228 | 89 | |
GenBank mRNA | AF076687.1 | - | 9 - 10 | 1222 | 89 | |
AF092840.1 | - | 9 - 10 | 1211 | 89 | ||
AK031933.1 | - | 9 - 10 | 1214 | 89 | ||
AK132302.1 | - | 9 - 10 | 1269 | 89 | ||
AK134960.1 | - | 9 - 10 | 1256 | 89 | ||
AK159790.1 | - | 9 - 10 | 1235 | 89 | ||
AK160614.1 | - | 9 - 10 | 1195 | 89 | ||
BC003719.1 | - | 9 - 10 | 1237 | 89 | ||
BC013190.1 | - | 9 - 10 | 1228 | 89 | ||
BC055061.1 | - | 9 - 10 | 1220 | 6528890 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
