Assay Details
Gene Symbol: Usp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013919.4 | NP_038947.2 | 7 - 8 | 1336 | 61 | |
GenBank mRNA | AF177759.1 | - | 6 - 7 | 1135 | 61 | |
AK011148.1 | - | 7 - 8 | 1346 | 61 | ||
AK013252.1 | - | 7 - 8 | 1331 | 61 | ||
BC021903.1 | - | 7 - 8 | 1326 | 3985792 | 61 | |
KF646669.1 | - | 6 - 7 | 854 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
