Assay Details
Gene Symbol: Usp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013919.4 | NP_038947.2 | 8 - 9 | 1442 | 90 | |
GenBank mRNA | AF177759.1 | - | 7 - 8 | 1241 | 90 | |
AK011148.1 | - | 8 - 9 | 1452 | 90 | ||
AK013252.1 | - | 8 - 9 | 1437 | 90 | ||
BC021903.1 | - | 8 - 9 | 1432 | 3985792 | 90 | |
KF646669.1 | - | 7 - 8 | 960 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
