Assay Details
Gene Symbol: Ngef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111314.1 | NP_001104784.1 | 5 - 6 | 1084 | 59 | |
NM_019867.2 | NP_063920.2 | 3 - 4 | 721 | 59 | ||
GenBank mRNA | AJ238898.1 | - | 3 - 4 | 702 | 59 | |
AK138433.1 | - | 3 - 4 | 690 | 59 | ||
AY038025.1 | - | 3 - 4 | 696 | 59 | ||
BC039279.1 | - | 5 - 6 | 1084 | 4973826 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
