Assay Details
Gene Symbol: Ngef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111314.1 | NP_001104784.1 | 6 - 7 | 1243 | 79 | |
NM_019867.2 | NP_063920.2 | 4 - 5 | 880 | 79 | ||
GenBank mRNA | AJ238898.1 | - | 4 - 5 | 861 | 79 | |
AK138433.1 | - | 4 - 5 | 849 | 79 | ||
AY038025.1 | - | 4 - 5 | 855 | 79 | ||
BC039279.1 | - | 6 - 7 | 1243 | 4973826 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
