Assay Details
Gene Symbol: Spats2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139140.1 | NP_631879.1 | 1 - 2 | 46 | 64 | |
GenBank mRNA | AB079132.1 | - | 1 - 2 | 46 | 64 | |
AK048720.1 | - | 1 - 2 | 88 | 64 | ||
AK159335.1 | - | 1 - 2 | 140 | 64 | ||
BC072579.1 | - | 1 - 2 | 313 | 64 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB079132.1,AK048720.1,NM_139140.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
