Assay Details
Gene Symbol: Ntrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008746.5 | NP_032772.3 | 6 - 7 | 822 | 58 | |
NM_182809.2 | NP_877961.1 | 6 - 7 | 822 | 58 | ||
GenBank mRNA | AF035399.1 | - | 7 - 8 | 917 | 58 | |
AF035400.1 | - | 6 - 7 | 971 | 58 | ||
AY336094.1 | - | 6 - 7 | 767 | 58 | ||
BC139764.1 | - | 7 - 8 | 834 | 40110345 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
