Assay Details
Gene Symbol: Ap1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301211.1 | NP_001288140.1 | 11 - 12 | 1474 | 73 | |
NM_009677.6 | NP_033807.2 | 12 - 13 | 1483 | 73 | ||
GenBank mRNA | AK034198.1 | - | 9 - 10 | 712 | 73 | |
AK036387.1 | - | 12 - 13 | 1426 | 73 | ||
AK145821.1 | - | 11 - 12 | 1405 | 73 | ||
BC006691.1 | - | 11 - 12 | 1150 | 73 | ||
BC052703.1 | - | 12 - 13 | 1356 | 5698075 | 73 | |
BC054535.1 | - | 12 - 13 | 1356 | 5698075 | 73 | |
X54424.1 | - | 11 - 12 | 1396 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
