Assay Details
Gene Symbol: Hal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010401.3 | NP_034531.1 | 9 - 10 | 1223 | 78 | |
GenBank mRNA | AK014518.1 | - | 9 - 10 | 1223 | 78 | |
AK028963.1 | - | 9 - 10 | 930 | 78 | ||
AK081409.1 | - | 9 - 10 | 925 | 78 | ||
BC057637.1 | - | 9 - 10 | 900 | 4211545 | 78 | |
BC157999.1 | - | 9 - 10 | 1205 | 9088147 | 78 | |
L07645.1 | - | 9 - 10 | 907 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
