Assay Details
Gene Symbol: Xpnpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133216.3 | NP_573479.3 | 16 - 17 | 1557 | 74 | |
GenBank mRNA | AF363970.1 | - | 16 - 17 | 1497 | 74 | |
AK052094.1 | - | 8 - 9 | 699 | 74 | ||
AK080423.1 | - | 5 - 6 | 305 | 74 | ||
AK146033.1 | - | 16 - 17 | 1537 | 74 | ||
AK149676.1 | - | 16 - 17 | 1496 | 74 | ||
AK158597.1 | - | 14 - 15 | 1515 | 74 | ||
AK166715.1 | - | 16 - 17 | 1457 | 74 | ||
AK169024.1 | - | 15 - 16 | 1383 | 74 | ||
BC021534.1 | - | 7 - 8 | 492 | 74 | ||
BC065174.1 | - | 16 - 17 | 1445 | 6415314 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
