Assay Details
Gene Symbol: Chrna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144803.2 | NP_659052.1 | 5 - 6 | 499 | 78 | |
GenBank mRNA | AY574261.1 | - | 4 - 5 | 383 | 78 | |
BC011490.1 | - | 5 - 6 | 499 | 4193582 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
