Assay Details
Gene Symbol: Oas1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145228.2 | NP_660263.1 | 5 - 6 | 1110 | 96 | |
GenBank mRNA | AB067530.1 | - | 5 - 6 | 1110 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149492 | NM_145228.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149492 | AB067530.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149492 | AK135899.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149492 | CF174513.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149492 | NM_001159934.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P313013 | NM_145228.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P313013 | AB067530.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P313013 | NM_145228.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P313013 | AB067530.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 486249 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75296 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 934277 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234791 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 762797 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 56745 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129381 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176470 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149492 | NM_145228.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149492 | AB067530.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149492 | AK135899.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149492 | CF174513.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149492 | NM_001159934.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P313013 | NM_145228.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P313013 | AB067530.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P313013 | NM_145228.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P313013 | AB067530.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041220 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041224 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041251 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041225 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041236 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041222 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041229 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041249 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041227 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041221 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041239 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041219 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041228 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041233 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041254 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041247 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041235 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041238 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041226 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041257 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176470 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129381 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05720314 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05720302 | - | - |
Whole Mouse Genome G4122A | A_52_P313013 | NM_145228.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P313013 | AB067530.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P313013 | NM_145228.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P313013 | AB067530.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P152602 | NM_145228.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P152602 | AB067530.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P152602 | CF174513.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P152602 | NM_001159934.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124104 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176470 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05720314 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129381 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04611869 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04611863 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04611881 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04611875 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04611864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041269 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041227 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04811081 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041271 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041263 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041251 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041222 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04480486 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041236 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041228 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04462768 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041238 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041233 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041242 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041239 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041272 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041278 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041221 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04683003 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041265 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041224 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041247 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041277 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041232 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041280 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041269 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041220 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041254 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041274 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041249 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041235 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10525176 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10525174 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10525175 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10525177 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10525178 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1425049_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10525173 | - | - |
Mouse Genome 430 2.0 Array | 1425049_at | - | - |
Mouse Genome 430 2.0 Array | 1425049_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 246729_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 246729_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500011789.mm.1 | TC0500001351.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500011789.mm.1 | NM_001159934.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500011789.mm.1 | NM_001159934.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500011789.mm.1 | NM_145228.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500011789.mm.1 | NM_145228.2 | Refseq |
Mouse Expression Set 430 | 1425049_at | - | - |
Mouse Expression Set 430 | 1425049_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1425049_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1425049_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425049_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425049_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 746825 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3360722 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003360722 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1242017 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1242017 | - | - |
mouse-6 v1.1 (Illumina) | scl27286.6.1_297-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3360722 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl27286.6.1_297-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3360722 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1242017 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1242017 | - | - |
