Assay Details
Gene Symbol: Kif1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153103.2 | NP_694743.2 | 20 - 21 | 2311 | 64 | |
GenBank mRNA | AB074017.1 | - | 18 - 19 | 1941 | 64 | |
AK046574.1 | - | 15 - 16 | 1552 | 64 | ||
AK173007.1 | - | 19 - 20 | 2018 | 64 | ||
BC006188.1 | - | 2 - 3 | 259 | 64 | ||
BC029286.1 | - | 20 - 21 | 2298 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
