Assay Details
Gene Symbol: Ago1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153403.2 | NP_700452.2 | 17 - 18 | 2261 | 99 | |
GenBank mRNA | AB081471.1 | - | 17 - 18 | 2261 | 99 | |
AK080954.1 | - | 15 - 16 | 2111 | 99 | ||
BC129914.1 | - | 17 - 18 | 2261 | 40111808 | 99 | |
BC129915.1 | - | 17 - 18 | 2257 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
