Assay Details
Gene Symbol: Gpsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199147.1 | NP_001186076.1 | 1 - 2 | 258 | 82 | |
GenBank mRNA | BC026486.1 | - | 1 - 2 | 249 | 82 | |
BC071197.1 | - | 1 - 2 | 255 | 6465025 | 82 | |
CK618701.1 | - | 1 - 2 | 258 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC071197.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
