Assay Details
Gene Symbol: Phldb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252442.1 | NP_001239371.1 | 14 - 15 | 3415 | 85 | |
NM_153412.4 | NP_700461.2 | 13 - 14 | 3256 | 85 | ||
GenBank mRNA | AF506821.1 | - | 13 - 14 | 3257 | 85 | |
AK035655.1 | - | 8 - 9 | 1019 | 85 | ||
AK050493.1 | - | 14 - 15 | 3423 | 85 | ||
BC050915.1 | - | 14 - 15 | 3271 | 6405379 | 85 | |
BC060683.1 | - | 14 - 15 | 3271 | 6405379 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
