Assay Details
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025392.1 | NP_001020563.1 | 9 - 10 | 2459 | 95 | |
NM_001025393.1 | NP_001020564.1 | 9 - 10 | 2459 | 95 | ||
NM_153787.2 | NP_722482.1 | 9 - 10 | 2453 | 95 | ||
GenBank mRNA | AK079110.1 | - | 5 - 6 | 709 | 95 | |
AK129071.1 | - | 6 - 7 | 1743 | 95 | ||
AK141834.1 | - | 9 - 10 | 2406 | 95 | ||
AK149724.1 | - | 9 - 10 | 2412 | 95 | ||
AK150017.1 | - | 9 - 10 | 2412 | 95 | ||
BC029682.1 | - | 9 - 10 | 2391 | 96 | ||
BC034300.1 | - | 9 - 10 | 2385 | 4507876 | 95 | |
BC086624.1 | - | 9 - 10 | 2441 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
