Assay Details
Gene Symbol: Rell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153793.2 | NP_722488.2 | 3 - 4 | 658 | 64 | |
GenBank mRNA | AK044097.1 | - | 3 - 4 | 621 | 64 | |
BC038500.1 | - | 3 - 4 | 601 | 4481989 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
