Assay Details
Gene Symbol: Esyt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028731.5 | NP_083007.2 | 9 - 10 | 1219 | 87 | |
GenBank mRNA | AK014813.1 | - | 9 - 10 | 1220 | 87 | |
AK079949.1 | - | 4 - 5 | 418 | 87 | ||
AK157352.1 | - | 9 - 10 | 1188 | 87 | ||
BC138937.1 | - | 9 - 10 | 1128 | 8861959 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK014813.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
