Assay Details
Gene Symbol: Esyt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014813.1 | - | 15 - 16 | 1650 | 122 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK014813.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
