Assay Details
Gene Symbol: Hook2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167991.1 | NP_001161463.1 | 1 - 2 | 164 | 111 | |
NM_133255.2 | NP_573518.2 | 1 - 2 | 164 | 111 | ||
GenBank mRNA | AK030599.1 | - | 1 - 2 | 157 | 111 | |
AK042048.1 | - | 1 - 2 | 152 | 111 | ||
AK136069.1 | - | 1 - 2 | 158 | 111 | ||
AK145768.1 | - | 1 - 2 | 132 | 111 | ||
AK161612.1 | - | 1 - 2 | 117 | 111 | ||
AK166954.1 | - | 1 - 2 | 144 | 111 | ||
AK169073.1 | - | 1 - 2 | 132 | 111 | ||
BC019486.1 | - | 1 - 2 | 140 | 4211990 | 111 | |
BC029967.1 | - | 1 - 2 | 105 | 111 | ||
BC055881.1 | - | 1 - 2 | 110 | 3590336 | 111 | |
BC080744.1 | - | 1 - 2 | 154 | 30471369 | 111 | |
CJ084759.1 | - | 1 - 2 | 193 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
