Assay Details
Gene Symbol: Evl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163395.1 | NP_001156867.1 | 10 - 11 | 1256 | 71 | |
NM_007965.3 | NP_031991.3 | 10 - 11 | 1504 | 71 | ||
GenBank mRNA | AK134563.1 | - | 10 - 11 | 1257 | 71 | |
BC059810.1 | - | 10 - 11 | 1504 | 6828523 | 71 | |
U72519.1 | - | 10 - 11 | 1267 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
