Assay Details
Gene Symbol: Mdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023397.4 | NP_075886.1 | 2 - 3 | 229 | 76 | |
GenBank mRNA | AF230273.1 | - | 2 - 3 | 95 | 76 | |
AK007319.1 | - | 2 - 3 | 213 | 76 | ||
AK160438.1 | - | 2 - 3 | 159 | 76 | ||
BC046613.1 | - | 2 - 3 | 152 | 6465642 | 76 | |
CJ176407.1 | - | 2 - 3 | 230 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
