Assay Details
Gene Symbol: Usp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024258.2 | NP_077220.2 | 5 - 6 | 603 | 156 | |
GenBank mRNA | AK004594.2 | - | 5 - 6 | 578 | 156 | |
AK160038.1 | - | 5 - 6 | 573 | 156 | ||
BC003278.1 | - | 5 - 6 | 532 | 3582180 | 156 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
