Assay Details
Gene Symbol: Ppp3r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024459.2 | NP_077779.2 | 4 - 5 | 523 | 114 | |
GenBank mRNA | AK133403.1 | - | 494 | 114 | ||
AK145541.1 | - | 4 - 5 | 500 | 114 | ||
AK160423.1 | - | 468 | 114 | |||
AK160938.1 | - | 4 - 5 | 509 | 114 | ||
AK163241.1 | - | 4 - 5 | 497 | 114 | ||
BC002267.1 | - | 2 - 3 | 351 | 114 | ||
S43864.1 | - | 3 - 4 | 281 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
