Assay Details
Gene Symbol: C1qtnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026161.3 | NP_080437.2 | 1 - 2 | 216 | 91 | |
GenBank mRNA | AK002948.1 | - | 1 - 2 | 145 | 91 | |
BC027315.1 | - | 1 - 2 | 64 | 91 | ||
CX243534.1 | - | 1 - 2 | 216 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK002948.1, NM_026161.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419451 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419449 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419481 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419483 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419478 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419479 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419476 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419450 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419480 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419448 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419482 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419477 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419484 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419475 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419445 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419446 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P633489 | DQ002397.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P633489 | AK004340.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P633489 | AK029777.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P633489 | AK002948.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P633489 | BC027315.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P633489 | DQ002397.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P633489 | AK004340.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P633489 | AK029777.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P633489 | AK002948.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P633489 | BC027315.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 391041 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811736 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175176 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P633489 | DQ002397.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P633489 | AK004340.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P633489 | AK029777.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P633489 | AK002948.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P633489 | BC027315.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P633489 | DQ002397.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P633489 | AK004340.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P633489 | AK029777.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P633489 | AK002948.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P633489 | BC027315.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419454 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419446 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419459 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419461 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419456 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419448 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419424 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419422 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419426 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419438 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419469 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419462 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419457 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419452 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419449 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419464 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419431 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419429 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419435 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419433 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419437 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419439 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419441 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419470 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419443 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21419466 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175176 | - | - |
Whole Mouse Genome G4122A | A_51_P488283 | BC087832.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P633489 | DQ002397.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P633489 | AK004340.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P633489 | AK029777.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P633489 | AK002948.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P633489 | BC027315.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P633489 | DQ002397.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P633489 | AK004340.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P633489 | AK029777.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P633489 | AK002948.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P633489 | BC027315.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131202 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175176 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02230549 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02230529 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02230531 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02230550 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02230528 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419449 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419478 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419479 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419450 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419476 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419484 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419483 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419482 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419481 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419445 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419475 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419448 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419477 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419446 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419451 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21419480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419489 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21419495 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419446 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419407 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419441 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419472 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419426 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02092356 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419418 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419464 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419406 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01610685 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419439 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419478 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01220219 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01610687 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419443 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02170584 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419448 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419434 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419459 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419410 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02254113 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419500 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02170586 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419462 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01630888 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02170585 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419488 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419476 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419437 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419435 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419466 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02092354 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02254122 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01246358 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01910455 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419473 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419433 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419492 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419497 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419495 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419452 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419456 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01435701 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419484 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01246360 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02170581 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419485 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419417 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419430 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419461 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419429 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419454 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419469 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01435703 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21419487 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10473753 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10473751 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10473752 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1441648_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417050_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10473750 | - | - |
Mouse Genome 430 2.0 Array | 1417050_at | - | - |
Mouse Genome 430 2.0 Array | 1441648_at | - | - |
Mouse Genome 430 2.0 Array | 1417050_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1441648_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67445_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67445_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200010385.mm.1 | TC0200001357.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200010385.mm.1 | NM_026161.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200010385.mm.1 | NM_026161.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200010388.mm.1 | TC0200001357.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200010388.mm.1 | NM_026161.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200010388.mm.1 | NM_026161.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200010384.mm.1 | TC0200001357.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200010384.mm.1 | NM_026161.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200010384.mm.1 | NM_026161.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200010387.mm.1 | TC0200001357.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200010387.mm.1 | NM_026161.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200010387.mm.1 | NM_026161.3 | Refseq |
Mouse Expression Set 430 | 1417050_PM_at | - | - |
Mouse Expression Set 430 | 1417050_at | - | - |
Mouse Expression Set 430 | 1441648_at | - | - |
Mouse Expression Set 430 | 1441648_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417050_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417050_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417050_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417050_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1441648_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1441648_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 927367 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 60619 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000060619 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1252584 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1252584 | - | - |
mouse-6 v1.1 (Illumina) | scl20653.2_227-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 60619 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl20653.2_227-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 60619 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1252584 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1252584 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl20653.2_227-S | - | - |
