Assay Details
Gene Symbol: Rad54l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030730.2 | NP_109655.2 | 5 - 6 | 883 | 67 | |
GenBank mRNA | AJ132389.1 | - | 5 - 6 | 784 | 67 | |
AK083903.1 | - | 5 - 6 | 864 | 67 | ||
AK143506.1 | - | 4 - 5 | 499 | 67 | ||
AK155151.1 | - | 5 - 6 | 840 | 67 | ||
BC133714.1 | - | 5 - 6 | 732 | 40143565 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
