Assay Details
Gene Symbol: Bicc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031397.2 | NP_113574.1 | 14 - 15 | 2039 | 73 | |
GenBank mRNA | AF319464.1 | - | 14 - 15 | 2040 | 73 | |
AK047271.1 | - | 4 - 5 | 1205 | 73 | ||
AK076154.1 | - | 14 - 15 | 2092 | 73 | ||
AK089066.1 | - | 14 - 15 | 1870 | 73 | ||
BC062174.1 | - | 6 - 7 | 908 | 73 | ||
BC111523.2 | - | 14 - 15 | 2040 | 40048106 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
