Assay Details
Gene Symbol: Srrt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109909.1 | NP_001103379.1 | 4 - 5 | 670 | 113 | |
NM_001109910.1 | NP_001103380.1 | 4 - 5 | 670 | 113 | ||
NM_031405.2 | NP_113582.1 | 4 - 5 | 670 | 113 | ||
GenBank mRNA | AK144300.1 | - | 4 - 5 | 661 | 113 | |
BC066831.1 | - | 4 - 5 | 629 | 30471339 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
