Assay Details
Gene Symbol: Arpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177616.1 | NP_001171087.1 | 9 - 10 | 1060 | 116 | |
NM_001177618.1 | NP_001171089.1 | 8 - 9 | 758 | 116 | ||
NM_001177619.1 | NP_001171090.1 | 9 - 10 | 1425 | 116 | ||
NM_033264.2 | NP_150289.1 | 8 - 9 | 893 | 116 | ||
GenBank mRNA | AF324451.1 | - | 8 - 9 | 853 | 116 | |
AK029467.1 | - | 9 - 10 | 1426 | 116 | ||
AK032417.1 | - | 9 - 10 | 986 | 116 | ||
AK032543.1 | - | 7 - 8 | 673 | 116 | ||
AK034376.1 | - | 7 - 8 | 606 | 116 | ||
AK153892.1 | - | 10 - 11 | 1709 | 116 | ||
BC053001.1 | - | 7 - 8 | 724 | 5686446 | 116 | |
BC098368.1 | - | 9 - 10 | 1007 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
