Assay Details
Gene Symbol: Cpsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164173.1 | NP_001157645.1 | 7 - 8 | 939 | 83 | |
NM_053193.2 | NP_444423.1 | 7 - 8 | 939 | 83 | ||
GenBank mRNA | AF322193.1 | - | 7 - 8 | 910 | 83 | |
AK155648.1 | - | 7 - 8 | 880 | 83 | ||
AK155806.1 | - | 7 - 8 | 861 | 83 | ||
BC011458.1 | - | 3 - 4 | 378 | 83 | ||
BC049941.1 | - | 7 - 8 | 761 | 83 | ||
BC056388.1 | - | 7 - 8 | 939 | 6854851 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
