Assay Details
Gene Symbol: Cpsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164173.1 | NP_001157645.1 | 31 - 32 | 3848 | 73 | |
NM_053193.2 | NP_444423.1 | 31 - 32 | 3848 | 73 | ||
GenBank mRNA | AF322193.1 | - | 31 - 32 | 3819 | 73 | |
AK155806.1 | - | 31 - 32 | 3770 | 73 | ||
BC011458.1 | - | 27 - 28 | 3297 | 73 | ||
BC049941.1 | - | 31 - 32 | 3670 | 73 | ||
BC056388.1 | - | 31 - 32 | 3848 | 6854851 | 73 | |
BC057945.1 | - | 16 - 17 | 2179 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
