Assay Details
Gene Symbol: Mta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054081.2 | NP_473422.2 | 14 - 15 | 1628 | 91 | |
GenBank mRNA | AB794093.1 | - | 15 - 16 | 1539 | 91 | |
AB794094.1 | - | 14 - 15 | 1488 | 91 | ||
AB794095.1 | - | 15 - 16 | 1492 | 91 | ||
AB794096.1 | - | 15 - 16 | 1539 | 91 | ||
AB794097.1 | - | 14 - 15 | 1441 | 91 | ||
AB794098.1 | - | 14 - 15 | 1488 | 91 | ||
AB794099.1 | - | 14 - 15 | 1488 | 91 | ||
AB794100.1 | - | 14 - 15 | 1441 | 91 | ||
AB794101.1 | - | 15 - 16 | 1539 | 91 | ||
AB794102.1 | - | 14 - 15 | 1428 | 91 | ||
AB794103.1 | - | 14 - 15 | 1488 | 91 | ||
AF288137.1 | - | 14 - 15 | 1590 | 91 | ||
AF463504.1 | - | 15 - 16 | 1539 | 91 | ||
BC005698.1 | - | 3 - 4 | 397 | 91 | ||
BC024527.1 | - | 7 - 8 | 854 | 91 | ||
BC025167.1 | - | 3 - 4 | 385 | 91 | ||
BC112910.1 | - | 15 - 16 | 1679 | 6515946 | 91 | |
CX203022.1 | - | 2 - 3 | 324 | 91 | ||
CX213009.1 | - | 1 - 2 | 73 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
