Assay Details
Gene Symbol: Arnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007488.3 | NP_031514.3 | 12 - 13 | 1467 | 58 | |
GenBank mRNA | AB093225.1 | - | 14 - 15 | 1647 | 58 | |
AK032628.1 | - | 12 - 13 | 1448 | 58 | ||
D63644.1 | - | 12 - 13 | 1343 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
