Assay Details
Gene Symbol: Anpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008486.2 | NP_032512.2 | 15 - 16 | 2349 | 67 | |
GenBank mRNA | AK143746.1 | - | 15 - 16 | 2292 | 67 | |
AK171427.1 | - | 16 - 17 | 2711 | 67 | ||
BC005431.1 | - | 15 - 16 | 2315 | 3593307 | 67 | |
BC017011.1 | - | 15 - 16 | 2373 | 3590117 | 67 | |
BC040792.1 | - | 15 - 16 | 2373 | 3590117 | 67 | |
U77083.1 | - | 15 - 16 | 2281 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
