Assay Details
Gene Symbol: Ppt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008917.3 | NP_032943.2 | 1 - 2 | 152 | 133 | |
GenBank mRNA | AF071025.1 | - | 1 - 2 | 169 | 133 | |
AF087568.1 | - | 1 - 2 | 130 | 133 | ||
AF326557.1 | - | 1 - 2 | 136 | 133 | ||
AF326558.1 | - | 1 - 2 | 136 | 133 | ||
AK014561.1 | - | 1 - 2 | 134 | 133 | ||
AK148314.1 | - | 1 - 2 | 127 | 133 | ||
AK151055.1 | - | 1 - 2 | 127 | 133 | ||
AK151107.1 | - | 1 - 2 | 138 | 133 | ||
AK152474.1 | - | 1 - 2 | 136 | 133 | ||
AK152940.1 | - | 1 - 2 | 138 | 133 | ||
AK153106.1 | - | 1 - 2 | 133 | 133 | ||
AK153316.1 | - | 1 - 2 | 133 | 133 | ||
AK171668.1 | - | 1 - 2 | 134 | 133 | ||
CB600397.1 | - | 1 - 2 | 155 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
