Assay Details
Gene Symbol: Ahrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009644.2 | NP_033774.1 | 6 - 7 | 655 | 76 | |
GenBank mRNA | AB015140.1 | - | 6 - 7 | 655 | 76 | |
AK154152.1 | - | 6 - 7 | 644 | 76 | ||
AK154761.1 | - | 6 - 7 | 677 | 76 | ||
AK155711.1 | - | 6 - 7 | 677 | 76 | ||
AK173134.1 | - | 3 - 4 | 297 | 76 | ||
BC118534.1 | - | 5 - 6 | 461 | 40095111 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
