Assay Details
Gene Symbol: Ahrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009644.2 | NP_033774.1 | 8 - 9 | 982 | 109 | |
GenBank mRNA | AB015140.1 | - | 8 - 9 | 982 | 109 | |
AK154152.1 | - | 8 - 9 | 971 | 109 | ||
AK154761.1 | - | 8 - 9 | 1004 | 109 | ||
AK155711.1 | - | 8 - 9 | 1004 | 109 | ||
BC118534.1 | - | 7 - 8 | 788 | 40095111 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
