Assay Details
Gene Symbol: Pde3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018779.1 | NP_061249.1 | 1 - 2 | 1276 | 62 | |
GenBank mRNA | AF099187.1 | - | 1 - 2 | 1276 | 62 | |
AK004638.1 | - | 1 - 2 | 581 | 62 | ||
AK041273.1 | - | 1 - 2 | 553 | 62 | ||
AK052374.1 | - | 1 - 2 | 554 | 62 | ||
AK164864.1 | - | 1 - 2 | 56 | 62 | ||
BC138265.1 | - | 1 - 2 | 1234 | 8861284 | 62 | |
BC138266.1 | - | 1 - 2 | 1234 | 8861285 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
