Assay Details
Gene Symbol: Ncoa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242558.1 | NP_001229487.1 | 3 - 4 | 534 | 82 | |
NM_019825.3 | NP_062799.2 | 3 - 4 | 534 | 82 | ||
GenBank mRNA | AF216186.1 | - | 2 - 3 | 425 | 82 | |
AK144338.1 | - | 5 - 6 | 773 | 82 | ||
AK171864.1 | - | 3 - 4 | 695 | 82 | ||
BC031113.1 | - | 3 - 4 | 538 | 82 | ||
BC083187.1 | - | 3 - 4 | 534 | 30544739 | 82 | |
JF707634.1 | - | 2 - 3 | 393 | 82 | ||
JF707635.1 | - | 2 - 3 | 393 | 82 | ||
JF707636.1 | - | 2 - 3 | 393 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
