Assay Details
Gene Symbol: Ncoa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242558.1 | NP_001229487.1 | 6 - 7 | 1687 | 73 | |
NM_019825.3 | NP_062799.2 | 6 - 7 | 1687 | 73 | ||
GenBank mRNA | AF216186.1 | - | 5 - 6 | 1572 | 73 | |
AK144338.1 | - | 8 - 9 | 1925 | 73 | ||
AK171864.1 | - | 6 - 7 | 1845 | 73 | ||
BC083187.1 | - | 6 - 7 | 1687 | 30544739 | 73 | |
JF707634.1 | - | 5 - 6 | 1546 | 73 | ||
JF707635.1 | - | 5 - 6 | 1546 | 73 | ||
JF707636.1 | - | 5 - 6 | 1546 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
