Assay Details
Gene Symbol: Ncoa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242558.1 | NP_001229487.1 | 7 - 8 | 1839 | 78 | |
NM_019825.3 | NP_062799.2 | 7 - 8 | 1839 | 78 | ||
GenBank mRNA | AF216186.1 | - | 6 - 7 | 1724 | 78 | |
AK144338.1 | - | 9 - 10 | 2077 | 78 | ||
BC083187.1 | - | 7 - 8 | 1839 | 30544739 | 78 | |
JF707634.1 | - | 6 - 7 | 1698 | 78 | ||
JF707635.1 | - | 6 - 7 | 1698 | 78 | ||
JF707636.1 | - | 6 - 7 | 1698 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
