Assay Details
Gene Symbol: Ncoa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242558.1 | NP_001229487.1 | 8 - 9 | 2955 | 106 | |
NM_019825.3 | NP_062799.2 | 8 - 9 | 2955 | 106 | ||
GenBank mRNA | AF135169.1 | - | 1 - 2 | 453 | 106 | |
AF216186.1 | - | 7 - 8 | 2840 | 106 | ||
AK129079.1 | - | 1 - 2 | 114 | 106 | ||
AK142566.1 | - | 2 - 3 | 1140 | 106 | ||
AK171864.1 | - | 8 - 9 | 3113 | 106 | ||
BC083187.1 | - | 8 - 9 | 2955 | 30544739 | 106 | |
JF707634.1 | - | 7 - 8 | 2814 | 106 | ||
JF707635.1 | - | 7 - 8 | 2814 | 106 | ||
JF707636.1 | - | 7 - 8 | 2814 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
