Assay Details
Gene Symbol: Gad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008078.2 | NP_032104.2 | 5 - 6 | 938 | 119 | |
GenBank mRNA | AF326549.1 | - | 5 - 6 | 602 | 119 | |
AF326550.1 | - | 5 - 6 | 602 | 119 | ||
AK136834.1 | - | 5 - 6 | 638 | 119 | ||
BC018380.1 | - | 5 - 6 | 653 | 4482097 | 119 | |
D42051.1 | - | 5 - 6 | 954 | 119 | ||
KC134211.1 | - | 5 - 6 | 602 | 119 | ||
L16980.1 | - | 5 - 6 | 674 | 119 | ||
S67454.1 | - | 1 - 2 | 80 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
