Assay Details
Gene Symbol: Max | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008558.2 | NP_032584.2 | 2 - 3 | 285 | 77 | |
GenBank mRNA | BC138671.1 | - | 2 - 3 | 141 | 8861692 | 77 |
BC138672.1 | - | 2 - 3 | 141 | 8861693 | 77 | |
M63903.1 | - | 2 - 3 | 222 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
