Assay Details
Gene Symbol: Mcm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008564.2 | NP_032590.2 | 14 - 15 | 2578 | 65 | |
GenBank mRNA | AK037751.1 | - | 14 - 15 | 2503 | 65 | |
AK077801.1 | - | 14 - 15 | 2486 | 65 | ||
AK088156.1 | - | 14 - 15 | 2491 | 65 | ||
AK088378.1 | - | 14 - 15 | 2478 | 65 | ||
AK129039.1 | - | 14 - 15 | 2459 | 65 | ||
AK145176.1 | - | 14 - 15 | 2490 | 65 | ||
AK146187.1 | - | 14 - 15 | 2493 | 65 | ||
AK146375.1 | - | 14 - 15 | 2502 | 65 | ||
BC055318.1 | - | 14 - 15 | 2487 | 5696348 | 65 | |
D86725.1 | - | 14 - 15 | 2498 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
